Z684635
Ace stirrer bearing, mechanical seal
for 10 mm shaft, #25 Ace-Thred
About This Item
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
Ace Glass 8050-12
diam.
10 mm
fitting
#25 Ace-Thred
General description
Equally good in both vacuum and pressure applications as low as -0.6 Torr. Rated for up to 400 rpm with both glass (polished rather than precision ground is best) and stainless steel shafts of 10, 19 and 28 mm diameter.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)