Key Documents
Z683760
Sigma-Aldrich
Aldrich® Molarmatic® measuring cylinders
cylinder set for acids, 1-3 M, Class A, calibrated to contain, volume 350 mL
Synonym(s):
graduated cylinders, graduated glass cylinders
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
glass
packaging
set of 1 (1 of each variety of the Molarmatic cylinders)
volume measuring range
350 mL
General description
The Molarmatic measuring cylinders are conveniently designed graduated cylinders that have a general-purpose 350 mL graduated scale on one side and a 1 to 3 Molar scale on the other for making standard acid solutions. These graduated cylinders eliminates repetitive Molar calculations and potential errors.
Just fill the cylinder to the desired Molar concentration mark with concentrated acid and pour the measured amount into 1 L water. The low profile cylinder shape is stable and fits easily into glassware storage cabinets. Cylinders are Class A with permanently etched graduations in 5.0 mL increments, calibrated "to contain".
Note: The set (Z683760) contains one of each acid measuring cylinder in a foam-lined plastic storage case.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
MOLARMATIC is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)