Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z682012

Scaffdex CellCeram insert scaffolds

ceramic scaffold, diam. × thickness 10 mm × 3 mm, sterile

Synonym(s):

3D, 3D Cell Culture, 3D scaffolds, cell scaffolds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107

sterility

sterile

manufacturer/tradename

Scaffdex CM00001

diam. × thickness

10 mm × 3 mm

color

white

General description

Scaffdex ceramic scaffold CellCeram is made of bioabsorbable hydroxyapatite and ß-tricalciumphosphate composite containing 60% hydroxyapatite and 40% ß-tricalciumphosphate. Scaffdex CellCeram is indicated for research purposes only, not for clinical use.

CellCeram has a foam type structure with average porosity of 83%. Average pore size is 200-400 μm with overall range of 100-800 μm. It also contains some microporosity and interconnected pores. CellCeram is available with diameter of 10 mm and height of 3 mm. It fits to 24- and 48 well plates. CellCrown24 and CellCrown48 can be used as assisting devices to maintain the firm position of CellCeram on the bottom of the well.

CellCeram is available as single packed and sterile, sterilized by gamma irradiation. CellCeram is for single use, only. It is recommended to put CellCeram scaffolds into cell culturing media one day before initiation of the cell culturing for the sufficient wetting of the scaffold.

Legal Information

CellCeram is a trademark of Scaffdex Oy

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service