Z681431
BriskHeat® temperature controller
AC/DC input 240 V AC, UK plug
About This Item
Recommended Products
description
Accuracy: +/- 0.6% of Span, +/- 1 Digit
manufacturer/tradename
BriskHeat
temperature measuring range
0-600 °C
32-999 °F
L × W × H
5.5 in. (140 mm) × 4.25 in. (108 mm) × 1.75 in. (45 mm)
Looking for similar products? Visit Product Comparison Guide
General description
- On-Off control
- Affordable and compact
- Compatible with a wide range of heaters and applications
- Easy programming
- Supplied with built-in J type thermocouple - 5 ft (1.5 m) factory fitted
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service