Key Documents
Z681172
Colored Microscope slides
yellow
About This Item
UNSPSC Code:
41122600
material
borosilicate glass 3.3
manufacturer/tradename
DURAN® 235502305
W × L × H
26 mm × 76 mm × 1.0 mm
color
yellow
binding type
non-treated surface
General description
These have one surface with a white, blue or yellow label field. Hence different preparations can be clearly differentiated by their label. The color printing of the label field is resistant against all solvents normally used in the laboratory.
These 90° ground white PRINT microscope slides with rounded corners are ideal for use in automatic machines, as glass breakages in the cassettes and on the conveyer belts of the machines can then be avoided.
  • Microscope slide in standard dimensions 26 x 76 mm
  • Glass thickness 1.0 ± 0.05 mm
  • The edges of the glass slides are uniform and crystal clear
  • Very uniform wettability
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)