Z681067
Duran® plastic-coated laboratory bottles
capacity 3,500 mL
Synonym(s):
218056903
About This Item
Recommended Products
material
borosilicate glass 3.3
round bottle
feature
without cap
packaging
pack of 1 ea
manufacturer/tradename
DURAN® 21 805 69 03
parameter
max. pressure (Not for use under pressure)
capacity
3,500 mL
diam. × H
160 mm × 295 mm
fitting
GL 45 neck thread
Looking for similar products? Visit Product Comparison Guide
General description
Please note: Cap not included.
Features and Benefits
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- The external polymer coating provides scratch, leak* and splinter* protection (* only applies to bottles 2000 ml and less)
- Highly resistanct, long service-life coating
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- UV protection up to approx. 380 nm due to external coating
- Complete with screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
- Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service