Z680141
BioCLAVE™ 16 digital autoclave
volume 16 L, AC/DC input 230 V AC, UK plug
About This Item
Recommended Products
feature
CE compliant
manufacturer/tradename
Benchmark Scientific B4000-16-UK Plug
chamber W × L
9 in. (23 cm) × 13.75 in. (35 cm)
external W × L × H
22 in. (56 cm) × 17.5 in. (44.5 cm) × 15.7 in. (40 cm)
trays W × L
6.5 in. (16.5 cm) × 10.75 in. (27 cm)
volume
16 L
16000 mL
Looking for similar products? Visit Product Comparison Guide
General description
Each program has a choice of two temperature settings:121 °C and 134 °C.
For special applications, requiring variations in cycle parameters, all settings on the BioClave 16 are fully adjustable with a digital program control.
Fitted with a mechanical and electrical safety interlock that prevents the door from being opened until the pressure has reached zero PSI.
Printer available separately.
Includes: 3 x loading trays, tray rack, tray handling tool, door adjustment tool, 2 x Draining hose, instruction manual, 2 x spare fuse power (20A), 2 x spare fuse for valve (3A), 2 x spare fuse for mainboard (1A), and door seal
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service