Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z679615

Hanna Checker® HC handheld colorimeter

Nitrite ULR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

General description

The Hanna Checker HC′s bridge the gap between simple chemical test kits and professional instrumentation. Chemical test kits are not very accurate and only give 5 to 10 points resolution while professional instrumentation can cost hundreds of dollars and can be time consuming to calibrate and maintain.

The Hanna Checker HC′s are accurate and affordable. The contoured style of these Checker HC′s fit in your palm and pocket perfectly and the large LCD is easy to read. The auto shut-off feature assures the battery life will not be drained if you forget to turn it off.

  • Easier to use and more accurate than chemical test kits
  • Large, easy to read digits
  • Auto shut-off
  • Dedicated to a single parameter
  • Uses 10 mL glass cuvettes
  • Use for quick and accurate on the spot analysis
  • Single button operation: Zero and Measure
  • Operated by a single AAA battery

Legal Information

Checker is a registered trademark of Hanna Instruments

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service