Key Documents
Z678732
Flamless butane heat tool kit
Master Ultratorch UT-40Si kit
About This Item
UNSPSC Code:
40101834
L
208 mm
General description
Use to pinpoint heat, remove surface mount components, repair vinyl, shrink heat-shrinkable tubing, solder & desolder, spot dry, strip insulation with hot knife.
  • Adjustable temperature lever for temps up to 1200 °F, equivalent to a 20-80 watt soldering iron
  • Compact size, lightweight and completely portable, features a rugged easy to hold plastic body
  • Convenient slide switch ignition system for quick ignition
  • UL & GS/TUV Listed for user safety
Components
Unit, solder tip, heat tip, hot knife tip, shrink attachment, cap, sponge & stand.
Choose from one of the most recent versions:
Certificates of Analysis (COA)