Key Documents
Z678511
Valve box for Gemini 88
pressure 30 psi
About This Item
UNSPSC Code:
41121500
manufacturer/tradename
KD Scientific 780389
parameter
30 psi pressure
General description
PTFE valve box with 2 x 3-way valves with 2 common open/close ports. In continuous operation the 2 common ports connect to the syringe tips, the other ports are plumbed in a cross over circuit.
For use with Gemini 88 syringe pump.
Choose from one of the most recent versions:
Certificates of Analysis (COA)