Z677906
Amicon Ultra 0.5 mL centrifugal filters
MWCO 100 kDa
About This Item
Recommended Products
material
polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing
sterility
non-sterile
manufacturer/tradename
Millipore UFC510024
tube size
1.5 mL
working volume
1.5 mL
pore size
100 kDa MWCO
General description
- Concentrates 500 μL down to 15 μL
- Concentration factor 25 x to 30 x
- High sample recovery (greater than 90% of dilute starting solution)
- Fast processing time - 10 to 30 minutes
- Consistent recoveries thanks to the reverse spin capability
- Reverse spin capability provides consistent recoveries
The Amicon Ultra-0.5 device is supplied with two microcentrifuge tubes. During operation, one tube is used to collect filtrate; the other tube is to recover the concentrated sample.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
How do I use the Amicon Ultra 0.5ML Centrifugal Filters?
The vendor instruction guide for the Amicon Ultra 0.5ML Centrifugal Filters is attached.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.