Key Documents
Z677779
BrassTec gloves
size XXL
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
(para-aramid, cotton, aluminium, silicone)
size
XXL
sterility
non-sterile
feature
latex free
powder-free
reusable
standard type EN388 (cat. II)
standard type EN407 (cat. III)
packaging
pack of 1 pair
manufacturer/tradename
KCL 947
L × thickness
360-380 mm × 3.10 mm
color
gray
General description
The aluminum-laminated hand back of the BrassTec glove protects the glove user up to a radiant heat of 1000°C (short-term). The 3-layer system also offers protection against conducted heat of 250°C while the silicone palm offers good mechanical protection.
Legal Information
BrassTec is a trademark of KCL GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)