Key Documents
Z677205
Dermatril LS nitrile gloves
size M
About This Item
UNSPSC Code:
46181504
material
blue nitrile
size
M
sterility
sterile
feature
CE compliant
manufacturer/tradename
KCL 747
L × thickness
280 mm × 0.11 mm
General description
  • Individually packed in pairs of sterile gloves
  • Antistatic
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Roughened fingertips for better grip
  • Rolled cuff edge for easier donning
  • CE compliant
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 61340-5-1, ASTM 1671:2001
Dermatril LS is a sterile, extended cuff, blue nitrile, lightly powdered glove providing a high level of protection against chemical and bacteriological contamination. The Dermatril LS disposable glove also displays very good physical durability in comparison to conventional disposable gloves.
All Dermatril LS disposable gloves are INDIVIDUALLY marked with the appropriate standards mark applied on the inside using a food-friendly dye.
  • Individually packed in pairs of sterile gloves
  • Antistatic
  • Free from allergenic substances such as thiurames, benzothiazoles and thiourea
  • Roughened fingertips for better grip
  • Rolled cuff edge for easier donning
  • CE compliant
  • Meet standards: EN 374 (cat. III), EN 388 (cat. II), EN 61340-5-1, ASTM 1671:2001
Legal Information
Dermatril is a trademark of KCL GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)