Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z677094

Millipore

Amicon Ultra 0.5 mL centrifugal filters

MWCO 3 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.24

material

polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing

sterility

non-sterile

manufacturer/tradename

Millipore UFC500324

tube size

1.5 mL

working volume

1.5 mL

pore size

3 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

With their vertical membrane design, Amicon Ultra 0.5 mL filters deliver great performance and lower spin times.

  • Concentrates 500 μL down to 15 μL
  • Concentration factor 25 x to 30 x
  • High sample recovery (greater than 90% of dilute starting solution)
  • Fast processing time - 10 to 30 minutes
  • Consistent recoveries thanks to the reverse spin capability
  • Reverse spin capability provides consistent recoveries

The Amicon Ultra-0.5 device is supplied with two microcentrifuge tubes. During operation, one tube is used to collect filtrate; the other tube is to recover the concentrated sample.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachael M Cunningham et al.
Journal of inorganic biochemistry, 189, 124-133 (2018-09-25)
Identifying the interactions of small molecules with biomolecules in complex cellular environments is a significant challenge. As one important example, despite being widely used for decades, much is still not understood regarding the cellular targets of Pt(II)-based anticancer drugs. In
Joseph S Bednash et al.
Cellular signalling, 79, 109859-109859 (2020-12-01)
The NACHT, LRR and PYD domains-containing protein 3 (NLRP3) inflammasome is a multimeric, cytoplasmic, protein complex that regulates maturation and secretion of interleukin (IL)-1β, a potent pro-inflammatory cytokine. Critical to host defense against pathogens, IL-1β amplifies early innate immune responses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service