Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z676829

ART® barrier tip

Empty reload rack, size , medium, for 20ul, B, 200ul and 300ul tips

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.01

sterility

non-sterile

feature

Empty reload rack
for 20ul, B, 200ul and 300ul tips

manufacturer/tradename

Thermo MBP 2006-ER

technique(s)

PCR: suitable

size

, medium

Looking for similar products? Visit Product Comparison Guide

General description

The self-sealing ART barrier tips are supplied in environmentally friendly packaging. The sterile, barrier tip reload system combines individually packaged blister tubs made of recycled plastic and new hinged racks. The push down blister tub easily inserts the ART barrier tips into the hinged racks providing stackable, space saving racks that offer easy, one-handed use. Empty hinged rack part numbers allow for easy conversion to the ART Reload System.

Manufactured according to the industry′s highest quality standards, ART tips are certified as RNase, DNase and Pyrogen free, and are ideal for the pipetting of radioactive samples, nucleic amplification procedures, or any applications where critical sample handling is required.

Available as reload inserts consisting of sterile pre-loaded blister tubs of 96 tips with or without hinged racks. The empty hinged racks are also available separately.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service