Key Documents
Z676373
Tabletop microtorch
stable tip-resistant design with easy-grip handle for safety
About This Item
UNSPSC Code:
40101834
NACRES:
NB.51
manufacturer/tradename
Master Appliance MT-30
H × W × D
5.1 in. × 3 in. × 3.5 in.
General description
The low profile, solid design of this portable benchtop burner provides a safe, portable flame in any work area. Easily lit with a push-button auto ignition with an adjustable flame and temperature up to 1300°C. Up to one hour run time with each refill.
  • Burner requires no wicks to replace or gas lines to install
  • Refills quickly from a standard butane fuel cartridge
  • 60 minutes continuous run time with a full charge
  • Stable tip-resistant design with easy-grip handle for safety
  • Flame temperature approximately 1300°C
  • Safety lock prevents unintentional ignition, locks for storage
Choose from one of the most recent versions:
Certificates of Analysis (COA)