Z675873
Talboys dual block heaters
AC/DC input 230 V AC, Schuko plug
About This Item
Recommended Products
material
aluminum plate
feature
CE compliant
manufacturer/tradename
Troemner 949513 EU
capacity
, holds 2 blocks
General description
Economical, versatile and compact. It accepts 2 separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes.
Each block has a thermometer well for measuring block temperature. Heats from ambient to 150 °C with two temperature adjustments, low range ambient to 100 °C/high range 75 °C to 150 °C. Takes only 50 mins to heat up to 100 °C.
The anodized aluminum modular heating blocks provide superior temperature stability and heat transfer.
- Compact and economical
- Holds 2 interchangeable heating blocks for maximum versatility
- Heating blocks sold separately
Choose from one of the most recent versions:
Certificates of Analysis (COA)