Key Documents
Z675407
Stuart melting point apparatus SMP30
AC/DC input 230 V AC
About This Item
UNSPSC Code:
41112203
NACRES:
NB.79
feature
CE compliant
manufacturer/tradename
Bibby
General description
The SMP30 can take three samples simultaneously within the optimised heating block and hold 8 results per tube in its memory. To allow maximum flexibility a plateau facility is included with variable ramp rate between 0.5 and 10°C in 0.1°C increments.
The tubes are illuminated with bright white LED′s to give the clearest view of the samples during the melt. The block has been designed for easy access for cleaning.
The SMP30 features the head up display, this unique feature displays a floating image of the block temperature, visible through the eyepiece, in front of the tubes.
  • Maximum temperature 400°C
  • Head up display
  • Integrated cooling 350°C to 50°C in 10 minutes
  • Large easy to read user friendly interface
Supplied with a pack of 100 melting point tubes, closed at both ends (Z673269).
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)