Key Documents
Z675261
Stuart SF1 accessories
2 × accessory side-arms and 4 × clamps
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
manufacturer/tradename
Bibby
W × depth × H
780 mm × 270 mm × 240 mm
weight
8.3 kg
General description
  • Vigorous shaking action
  • Timed operation or continuous running
  • Robust construction
  • Ideal for extractions
  • Speed range 8 to 800 oscillations/min
  • Amplitude 1.5 mm
Valuable time can be taken up mixing bottles and flasks by mixing by hand. Let the SF1 take the strain. Holding up to eight flasks or bottles, up to 500ml capacity, it creates a vigorous mixing action by simulating hand shaking. It is especially useful for applications where prolonged shaking is required as it will not get tired and can be left on continuously!
Robust construction, mounted on four rubber feet to absorb vibration and prevent unnecessary movement on the bench. Analogue timer covers 10 to 60 minutes with a manual override. Electronic feed-back control ensures a constant speed irrespective of load.
Supplied with two side arms, eight clamps and Allen key. An optional extension kit is available, catalogue number SF1/2 which consists of two extension arms and four extra clamps. It increases the capacity to twelve bottles or flasks.
Allows shaker to hold up to 12 vessels
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)