Key Documents
Z675245
Stuart SF1 flask shaker
AC/DC input 230 V AC
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
feature
CE compliant
packaging
pack of 1
manufacturer/tradename
Bibby
W × depth × H
780 mm × 270 mm × 240 mm
weight
8.3 kg
General description
  • Vigorous shaking action
  • Timed operation or continuous running
  • Robust construction
  • Ideal for extractions
  • Speed range 8 to 800 oscillations/min
  • Amplitude 1.5 mm
Valuable time can be lost when mixing bottles and flasks by hand. Let the SF1 take the strain. Holding up to eight flasks or bottles, up to 500 mL capacity, it creates a vigorous mixing action by simulating hand shaking. It is especially useful for applications where prolonged shaking is required as it will not get tired and can be left on continuously!
Robust construction, mounted on four rubber feet to absorb vibration and prevent unnecessary movement on the bench. Analogue timer covers 10 to 60 minutes with a manual override. Electronic feed-back control ensures a constant speed irrespective of load.
Supplied with two side arms, eight clamps and an Allen key. An optional extension kit is available, catalogue number SF1/2 which consists of two extension arms and four extra clamps. It increases the capacity to twelve bottles or flasks.
Comes with both Schuko plug and UK plug.
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)