Key Documents
Z675164
Stuart SB2 fixed speed rotator
AC/DC input 230 V AC
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
feature
CE compliant
manufacturer/tradename
Bibby
parameter
20 rpm speed
W × D × H
200 mm × 270 mm × 230 mm
weight
3.2 kg
General description
  • Ideal for keeping biological samples such as blood in suspension
  • Quick and easy loading and removal of tube
  • Choice of fixed or variable speed
  • Fully adjustable mixing angle
  • Accessory tube holders to hold a number of different sized tubes
  • Possible to hold two tube holders concurrently with new dual tube holder adaptor SB3/dual
  • Holder must be ordered separately
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)