Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z674966

Microflex® Synetron® Plus latex extended cuff examination gloves

size XL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504

material

natural gloves
natural natural rubber

size

XL

sterility

non-sterile

feature

CE compliant
disposable
latex free: no
powder-free

packaging

pack of 1 ea

manufacturer/tradename

Microflex SYP-INT-XL

availability

not available in USA and Canada

L × thickness

11.4 in. (290 mm) × 9.1 mil (0.23 mm)

Looking for similar products? Visit Product Comparison Guide

General description

Synetron Plus latex examination gloves are a strong barrier against many chemicals and biohazards, and its comfortable during extended periods of wear and is textured for a secure grip. The polymer coating ensures product consistency and easy donning.

  • Polymer-coating system provides effective donning in wet, damp or dry conditions
  • Extended cuff for increased protection
  • Made from soft natural rubber latex for improved elasticity and greater comfort
  • Reduced protein rating

Gloves comply with personal protective equipment directive 89/686/eec, complex design category; European medical device directive 93/42/EEC, CE class 1; type tested to EN 374-2, EN 374-3, EN 388 & EN 420, and also complies with en 455 parts 1, 2, & 3.

Thickness:
  • Cuff: 0.17 mm (6.7 mil)
  • Palm: 0.23 mm (9.1 mil)
  • Finger: 0.30 mm (11.8 mil)
  • Length: 290 mm (11.4")

Legal Information

Microflex is a registered trademark of CIT Healthcare LLC
Synetron is a registered trademark of CIT Healthcare LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service