Key Documents
Z674664
Slide spinner for slide drying
AC/DC input 110 V AC
About This Item
UNSPSC Code:
41100000
NACRES:
MA.01
manufacturer/tradename
Labnet C1303-T
availability
not available in EU
parameter
~4800 rpm speed
W × D × H
5 in. (12.7 cm) × 5 in. (12.7 cm) × 4.4 in. (11.2 cm)
General description
Simply insert your slides into the cassettes and close the lid. This Slide Spinner quickly accelerated to 4800 RPM, the optimal speed for quick drying without any splashing. Open the lid and the rotor comes to a stop and the dry slides can be removed. The cassettes catch all of the liquid, and can easily be removed, and the liquid can be poured out.
  • Quickly dries 2 slides
  • Ideal for removing wash buffer from microarrays prior to scanning
Removeable cassettes conveniently catch all liquid
Choose from one of the most recent versions:
Certificates of Analysis (COA)