Key Documents
Z674559
GyroMini nutating mixer
AC/DC input 110 V AC
Synonym(s):
lab equipment accessory
About This Item
UNSPSC Code:
41103801
NACRES:
NB.73
feature
CSA approved
manufacturer/tradename
Labnet S0500
availability
not available in EU
General description
  • Small foot print means less bench space needed
  • Gyratory action provides gentle, thorough mixing
  • Ideal for mixing blood samples in centrifuge tubes
  • Fixed speed and pitch provide optimum agitation
  • Safe for cold room use
The GyroMini is supplied with a dimpled mat for holding tubes firmly in place.
Legal Information
GyroMini is a trademark of Labnet Intl., Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)