Key Documents
Z674451
Duran® DG safety cap accessories
DG waste cap GL 45
About This Item
UNSPSC Code:
41121800
manufacturer/tradename
DURAN® 1168418
General description
Complete with GL 45 screw cap, connection screws and conical nipple (I.D. 3.2 mm, PTFE).
DG safety waste caps are ideal for safely collecting and storing liquid laboratory waste. With a choice of different connections and activated carbon filters, they increase safety for laboratory technicians.
  • Drip-free collection of used or contaminated media
  • Charcoal filters prevent harmful gas emissions
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)