Key Documents
Z674443
Duran® DG safety caps
3-ports with stopcock
Synonym(s):
dg cap, dg caps
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
manufacturer/tradename
DURAN® 11 684 16
General description
DG safety caps are high-grade PTFE, flexible connection systems which are designed specifically for handling aggressive, gas-forming liquids. Available with 1, 2 or 3 connection ports with stopcocks. Complete with 3.2 mm v-ring seal. GL 45 thread.
  • No dangerous gas emissions
  • No slipped tubes
  • No loss due to evaporation
  • Clean working environment
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)