Key Documents
Z673897
Hanna HI 9125 portable waterproof pH/ORP meter
About This Item
UNSPSC Code:
41115603
manufacturer/tradename
Hanna HI 9125
mV measuring range
±1999 mV, accuracy: 1 mV
±699 mV, accuracy: 0.2 mV
pH measuring range
-2.00-16.00, accuracy: 0.01
temperature measuring range
-9.9-120 °C, accuracy: ±0.5 °C
General description
For UK power adapter please order Z673978 for Hanna UK cord HI 710012
The HI 9125 is a heavy-duty waterproof pH/ORP meter designed with the latest microprocessor technology to provide laboratory results and accuracy under harsh industrial conditions.
The multi-level LCD displays pH and temperature readings simultaneously, while also displaying the electrode status and operational guides.
Battery Error Prevention Sysytem (BEPS) automatically turns the instrument off when battery level is too low to ensure reliable readings.
The HI 9125 can also be used to measure ORP in the mV range with 0.1 mV resolution.
  • Large multi-level display LCD
  • On-screen operational guide
  • Waterproof
  • Auto calibration at 1 or 2 points
Supplied with HI 1230B combination double junction, gel-filled pH electrode, HI 7662 SS temperature probe, electrode holder, pH 4.01 & 7.01 buffer solutions, 100 mL plastic beaker, 4 x 1.2 V AAA rechargeable batteries (up to 2000 hours life), inductive recharger, instruction manual and rugged carrying case.
These units are compatible with a wide range of pH electrodes with BNC connectors.
Choose from one of the most recent versions:
Certificates of Analysis (COA)