manufacturer/tradename
Hanna HI9124N
pH measuring range
-2.00-16.00, accuracy: 0.01
temperature measuring range
-9.9-120 °C, accuracy: ±0.5 °C
General description
The multi-level LCD displays pH and temperature readings simultaneously, while also displaying the electrode status and operational guides.
Battery Error Prevention System (BEPS) automatically turns the instrument off when battery level is too low to ensure reliable readings.
- Large multi-level display LCD
- On-screen operational guide
- Waterproof
- Auto calibration at 1 or 2 points
- Battery Error Prevention System (BEPS)
Supplied with HI 1230B combination double junction, gel-filled pH electrode, HI 7662 SS temperature probe, electrode holder, pH 4.01 & 7.01 buffer solutions, 100 mL plastic beaker, 4 x 1.2v AAA rechargeable batteries (up to 2,000 hours life), inductive recharger, instruction manual and rugged carrying case.
These units are compatible with a wide range of pH electrodes with BNC connectors.
Choose from one of the most recent versions:
Certificates of Analysis (COA)