Key Documents
Z673730
PROHEAT® VARITEMP® heat guns
PH-220-A6, AC/DC input 220 V AC, UK plug
About This Item
UNSPSC Code:
41102404
NACRES:
NB.76
material
plastic
feature
CE compliant
parameter
453 L/min flow rate
453 L/min temp. range
General description
These heat guns offer all the features of the PROHEAT heat guns, plus variable temperature control. Dial in any temperature from 130°F (49°C) to 840°F (449°C). Ideal for confined spaces or for applications where its lightweight construction can prevent operator fatigue.
Legal Information
PROHEAT is a registered trademark of Master Appliance Corp.
VARITEMP is a registered trademark of Master Appliance Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)