Key Documents
Z673722
PROHEAT® heat gun
PH-2100-A6, AC/DC input 230 V AC, UK plug
About This Item
UNSPSC Code:
41102404
NACRES:
NB.76
material
plastic
feature
CE compliant
parameter
260-538 °C temp. range
453 L/min flow rate
General description
Compact, lightweight heat guns with dual temperature settings of 430°F and 840°F (221°C and 449°C). Excellent source of flameless heat with air flow at 15 cfm. The PROHEAT heat guns meet most general laboratory applications.
Legal Information
PROHEAT is a registered trademark of Master Appliance Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)