Z673641
Talboys block heater
AC/DC input 110 V AC, US 2-pin plug
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NB.73
material
aluminum plate
manufacturer/tradename
Troemner 94951X
availability
not available in EU
capacity
, holds 1 block
General description
- Each block has a thermometer well for measuring block temperature
- Heats from ambient to 150°C with two temperature adjustments, low range ambient to 100°C/high range 75°C to 150°C.
- Heating blocks are sold sperately
This multi-purpose unit is ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures.
Economical, versatile and compact. It accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials,microplates, and PCR strips or tubes.
Each block has a thermometer well for measuring block temperature. Heats from ambient to 150°C with two temperature adjustments, low range ambient to 100°C/high range 75°C to 150°C.
The anodized aluminum modular heating blocks provide superior temperature stability and heat transfer.
Heating blocks are sold sperately.
Economical, versatile and compact. It accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials,microplates, and PCR strips or tubes.
Each block has a thermometer well for measuring block temperature. Heats from ambient to 150°C with two temperature adjustments, low range ambient to 100°C/high range 75°C to 150°C.
The anodized aluminum modular heating blocks provide superior temperature stability and heat transfer.
Heating blocks are sold sperately.
Features and Benefits
- Compact design takes up little bench space
- Interchangeable heating blocks for maximum versatility
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)