Key Documents
Z673331
Drilled blocks for Talboys block heater
titer plate
About This Item
UNSPSC Code:
41102404
NACRES:
NB.73
feature
wells 1
Holds 1 x plate
manufacturer/tradename
Troemner 949115
block L × W × H
3.75 in. (9.5 cm) × 3 in. (7.6 cm) × 2 in. (5.1 cm)
well D
13.5 mm
General description
Constructed from a solid anodized aluminum block, the close contact of tubes to block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Units are designed as a dry bath/incubator.
This is a double size block which fits dual block heater, Z675873.
Caution
To avoid possible electrical hazard, do not fill well or block with water or other liquids.
Choose from one of the most recent versions:
Certificates of Analysis (COA)