Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z673323

Drilled blocks for Talboys block heater

Holds 20 x 1.5 mL tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
NACRES:
NB.73

feature

Holds 20 x 1.5 mL tubes

manufacturer/tradename

Troemner 949110

block L × W × H

3.75 in. (9.5 cm) × 3 in. (7.6 cm) × 2 in. (5.1 cm)

well diam. × D

11.1 mm × 39.1 mm

Looking for similar products? Visit Product Comparison Guide

General description

Constructed from a solid anodized aluminum block, the close contact of tubes to block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature.
Units are designed as a dry bath/incubator.

Caution

To avoid possible electrical hazard, do not fill well or block with water or other liquids.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service