Z671967
KNF LIQUIPORT chemically-resistant diaphragm liquid pumps
AC/DC input 230 V AC, PTFE (head), flow rate 0.5-3.0 L/min
About This Item
Recommended Products
material
PTFE (head)
manufacturer/tradename
KNF NF 300 FT.18 S
parameter
0.5-3.0 L/min flow rate
tube I.D.
12 mm
weight
1.5 kg
output
24 watts (motor)
General description
The NF100 and NF300 pumps are available in either polypropylene or PTFE head materials, with FFPM valve. You should always check corrosion compatibility charts with the medium you intend to pump.
- 10 mWg pressure head
- 3 mWg suction head
- IP65 protection class
- Maintenance-free
- Self priming
- Dry running
- Adjustable flow rate
Choose from one of the most recent versions:
Certificates of Analysis (COA)