Z671894
KNF LIQUIPORT chemically-resistant diaphragm liquid pumps
AC/DC input 115 V AC, polypropylene (head), flow rate 0.2-1.3 L/min
About This Item
Recommended Products
material
polypropylene (head)
packaging
pack of 1 ea
manufacturer/tradename
KNF NF 100 KT.18 S
availability
not available in EU
parameter
0.2-1.3 L/min flow rate
tube I.D.
8 mm
weight
1 kg
output
12 watts (motor)
General description
The NF100 and NF300 pumps are available in either polypropylene or PTFE head materials, with FFPM valve. You should always check corrosion compatibility charts with the medium you intend to pump.
- 10 mWg pressure head
- 3 mWg suction head
- IP65 protection class
- Maintenance-free
- Self priming
- Dry running
- Adjustable flow rate
Choose from one of the most recent versions:
Certificates of Analysis (COA)