Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z671010

Whatman® Westran® PVDF membranes

Westran S, 0.2 μm PVDF blotting membrane, 10 x 10 cm sheet, 10/pk

Synonym(s):

Western blotting membranes, blotting membranes, protein blotting membranes, western blot sandwich

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

packaging

pack of 10

manufacturer/tradename

Whatman 10413052
Whatman Article No. 28415328 (US reference)

size

10 cm × 10 cm

pore size

0.2 μm

suitability

suitable for (Western blotting, protein sequencing and protein dot-blotting applications)

Looking for similar products? Visit Product Comparison Guide

General description

  • Protein binding capacity (over 200 μg/cm2) for easy signal detection
  • Chemical resistance needed for N-terminal sequencing
  • High protein retention even after harsh wash steps
  • Maximum capture of proteins during transfers minimizing sample loss
  • 0.2 μm pore size for higher surface area, resulting in better binding of low molecular weight proteins
  • Compatible for use with Western blotting applications
  • Available in popular pre-cut sizes for your application
  • Protein binding ability over 125 μg/cm2
  • Extremely low backgrounds with chemiluminescent and colorimetric applications providing you with clear signals and sharp bands
  • Excellent results with general protein stains such as Coomassie Brilliant Blue, Amido Black and Ponceau S Red
  • Increased strength allows for multiple stripping and reprobing, which results in savings

Westran Blotting Sandwiches

A pre-cut PVDF membrane and 2 sheets of 3MM Chr blotting paper are prepackaged into a sandwich to help save time. Sandwiches are available with either Westran S or CS.

Westran® PVDF Membranes


Whatman Westran PVDF (Polyvinylidine Fluoride) membranes are available in two formats: the Westran S used for protein sequencing and the Westran Clear Signal used for Western blotting.

Westran S

Westran S PVDF is a 0.2 μm pore size hydrophobic membrane designed specifically for protein sequencing applications. The small pore size of this membrane eliminates ‘blow-through′ and increases protein binding over a wide range of molecular weights.

Westran Clear Signal
Westran Clear Signal (CS) PVDF is a 0.45 μm membrane specifically designed for Western blotting and protein dot-blotting applications.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Westran is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanli Ma et al.
PloS one, 7(10), e47961-e47961 (2012-11-01)
Aflatoxin B1 (AFB1) is a potent carcinogen that can induce hepatocellular carcinoma. AFB1-8,9-exo-epoxide, one of AFB1 metabolites, acts as a mutagen to react with DNA and induce gene mutations, including the tumor suppressor p53. In addition, AFB1 reportedly stimulates IGF

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service