Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z670596

Whatman® Optitran® reinforced nitrocellulose membranes

Rolls - BA-S 85, pore size 0.45 μm, W × L 20 cm × 3 m

Synonym(s):

blotting membrane, nylon membrane, paper filtration

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Whatman 10439194
Whatman Article No. 28415489 (US reference)

W × L

20 cm × 3 m

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

Optitran membrane consists of pure 100% nitrocellulose supported by an inert polyester non-woven material within the membrane. The support does not affect transfer conditions or results and gives the membrane exceptional handling characteristics, allowing it to be reprobed repeatedly.

The Optitran nitrocellulose membrane provides high sensitivity with very low non-specific binding. Using standard nitrocellulose protocols, stringent washing and blocking conditions are not necessary.

The combination of flexibility, strength and excellent signal-to-noise ratios makes the Optitran membrane ideal, especially when experiments involve repeated stripping and reprobing. Optitran supported nitrocellulose membranes combine sensitivity, strength and savings. Optitran binding capacity: 75-90 μg/cm2; autoclavable (liquid cool cycle).

These provide the same excellent characteristics as the Protran nitrocellulose with the addition of handling strength, 75-90 μg/cm2 binding capacity.
  • High binding, low background
  • Superior handling characteristics
  • Triton free matrix

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Optitran is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva

pictograms

Flame

signalword

Danger

hcodes

pcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunniva Stordal Bjørklund et al.
BMC cancer, 15, 524-524 (2015-07-18)
Alternate transcripts from a single gene locus greatly enhance the combinatorial flexibility of the human transcriptome. Different patterns of exon usage have been observed when comparing normal tissue to cancers, suggesting that variant transcripts may play a role in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service