Key Documents
Z665711
Ohaus® Discovery semi-micro analytical balance
DV114C, AC/DC input 230 V AC
Synonym(s):
analytical scales, laboratory balances, weighing balances
About This Item
UNSPSC Code:
41111502
material
stainless steel weighing plate
feature
CE compliant
CSA approved
manufacturer/tradename
DV114C
weighing capacity measuring range
110 g, resolution: 0.1 mg
platform diam.
90 mm
General description
Ohaus Discovery series of semi-micro and analytical balances combine unmatched weighing performance with Ohaus′ innovative SmarText software, making it extremely reliable and easy-to-use. The Discovery features durable glass and steel construction and together with advanced AutoCal internal calibration makes it the best performing balance in its class.
Discovery models have capacities upto 310g and are perfect for university research, pharmaceutical research and drug discovery, and general lab applications.
Features and Benefits
  • Highest performance
  • Multiple weighing units
  • Environmental settings
  • Easy to set up and use
  • Advanced AutoCal calibration
  • Four methods of density determination
  • RRS232 interface with full GLP/GMP Protocol
  • Protective in-use cover
  • Up-front level indicator
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)