Key Documents
Z660639
Catalyst heater for Plas-Labs temperature and humidity box
AC/DC input 240 V AC, UK plug
Synonym(s):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
Select a Size
1 EA
$2,710.00
$2,710.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
1 EA
$2,710.00
About This Item
UNSPSC Code:
41103406
NACRES:
NB.35
$2,710.00
Check Cart for Availability
Notify Me
Get notified when this item is ready to ship via email.
manufacturer/tradename
Plas-Labs 800-HEATER
General description
The Plas-Labs catalyst heater has two primary functions for use in the Plas-Labs temperature and humidity glove box. The catalyst heater reduces trace amounts of oxygen and maintains incubation temperature levels of 35-37 °C. The internal atmosphere is drawn off the floor and up across heating elements and aluminized palladium pellets. Any oxygen present will react with your anaerobic gas mixture and be reduced to water vapor. When used in conjunction with the Plas-Labs temperature and humidity glove boxes with drying train attached, the vapor is then removed from the box as needed. CE compliant.
Legal Information
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)