Key Documents
Z655597
Hanna combination ISE electrodes
Silver/Sulfide, Solid-state
About This Item
UNSPSC Code:
41115603
material
polyetherimide body
description
Silver/Sulfide, Solid-state
manufacturer/tradename
Hanna HI 4115
ISE measuring range
107900-0.01 ppm
pH measuring range
2-8
parameter
0-80 °C temp. range
electrode immersion depth × O.D.
120 mm × 12 mm
General description
Determination of sulfide ions in waters, paper liquors, natural waters and soils.
These include a sensor and reference electrode in a single electrode body. Hanna combination ion selective electrodes provide selectivity and include superior double junction reference into an electrode body. Combination solid-state electrodes have a built-in solid-state sensor and quick refill reference electrode.
Hanna′s liquid membrane and fluoride combination electrodes have replaceable module construction and the Hanna double junction refence stability.
Choose from one of the most recent versions:
Certificates of Analysis (COA)