Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z655449

Hanna combination ISE electrodes

Bromide, Solid-state

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

material

polyetherimide body

manufacturer/tradename

Hanna HI 4102

ISE measuring range

79910-0.07991 ppm

pH measuring range

2-12.5

parameter

0-80 °C temp. range

electrode immersion depth × O.D.

120 mm × 12 mm

Looking for similar products? Visit Product Comparison Guide

General description

These include a sensor and reference electrode in a single electrode body. Hanna combination ion selective electrodes provide selectivity and include superior double junction reference into an electrode body. Combination solid-state electrodes have a built-in solid-state sensor and quick refill reference electrode.

Hanna′s liquid membrane and fluoride combination electrodes have replaceable module construction and the Hanna double junction refence stability.

Application

Determination of free bromide ions in emulsified food products, beverages, plants, soils and as an indicator for titration.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service