Key Documents
Z655406
Hanna specific analysis pH electrode
pH 0-12, temp. range -5-50 °C (23-122 °F)
About This Item
UNSPSC Code:
41115603
manufacturer/tradename
Hanna
pH measuring range
0-12
parameter
-5-50 °C temp. range (23-122 °F)
0.1 bar max. pressure
General description
pH electrode for skin and flat surfaces. Flat tipped glass body pH electrode with open junction and BNC connector for special applications.
Ideal for:
  • Skin
  • Leather
  • Paper
  • Emulsions
Specifications:
  • Single, Ag/AgCI
  • Open junction
  • Electrolyte viscolene
  • Glass body
Supplied fitted with:
  • BNC connector
  • 1m cable (coaxial)
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)