Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z655333

Hanna bench pH/ISE meter HI 4222

pH -2.0-20, accuracy: 0.1-0.002, ISE 1 × 10-7-9.99 × 1010, accuracy: 0.5%, temperature -20-120 °C, accuracy: 0.2 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

manufacturer/tradename

Hanna

ISE measuring range

1 × 10-7-9.99 × 1010, accuracy: 0.5%

mV measuring range

±2000, accuracy: 0.2

pH measuring range

-2.0-20, accuracy: 0.1-0.002

temperature measuring range

-20-120 °C, accuracy: 0.2 °C

size

159 mm × 230 mm × 93 mm

weight

800 g

General description

  • Exclusive Hanna CAL CHECK, five point calibration with standard and custom buffers
  • Direct calibration and measurement in multiple units
  • Incremental methods: Known addition, Known subtraction, Analyte addition, Analyte subtraction
  • 2pH/mV/ISE input channels
  • Calibration check: pH electrode and buffer condition
  • Auto endpoint
  • GLP - offset, slope, date, buffers used for calibration at push of a button
  • Real time graphing
  • USB & RS232 interface
Supplied in kit form, glass body pH electrode, temperature probe, 12 V DC adapter, pH 4 & 7 buffer solutions, electrode refilling solution, electrode holder and instructions.

Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service