Key Documents
Z655082
Carbolite® tube furnace end seals
1x NW16 flange, diam. ~32 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
manufacturer/tradename
Carbolite TS-032-042-0030
packaging
1 ea of
diam.
~32 mm
General description
Work tube end seals are required to contain a modified atmosphere and for working with vacuum, vacuum levels of 10 - 6 mbar are possible. End seals are manufactured from stainless steel and are for use with extended work tubes only.
End seal includes a clamp, seal plate, ′O′ ring seal, seal sleeve, clamp seal and an end plate with an NW16 flange.
Legal Information
Carbolite is a registered trademark of Carbolite Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)