Key Documents
Z655066
Carbolite® tube furnace end plugs
radiation shield, pack of 1
About This Item
UNSPSC Code:
41121800
NACRES:
NB.24
packaging
pack of 1
manufacturer/tradename
Carbolite RS-025-12C-001E
General description
For use on horizontal tube furnaces up to 1200°C only. Fits tubes up to 80 mm I.D.
For use with horizontal tube furnaces up to 1200C, with up to an 80mm ID process tube. Radiation shields effectively reduce heat loss out of the end of the process tube by combining a series of heat reflecting alloy discs on an alloy support tube. The result is an extended uniform temperature zone within the furnace work tube. A radiation sheild assembly is positioned inside of the process tube at either end of the furnace hot zone.
Legal Information
Carbolite is a registered trademark of Carbolite Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)