Key Documents
Z654515
Stuart Gyratory rocker, SSL3
load ~10 kg
About This Item
UNSPSC Code:
41100000
NACRES:
NB.73
Product Z654515 is not currently sold in your country. Contact Technical Service
AC/DC input
230 V AC
feature
CE compliant
manufacturer/tradename
Bibby
availability
available only in EU
parameter
5-70 rpm speed
~10 kg load
platform W × L
355 mm × 355 mm
General description
  • 3D gyratory motion, ideal for low foaming agitation, DNA extractions, staining and de-staining procedures
  • Angle of the tilt can be moved to any position by hand to optimize mixing vessels
  • Large platform rocker - ideal for multiple users
  • Gentle rocking action
  • Digital speed control and built in timer
  • Optional tier system available to increase capacity
Rockers are often used in conjunction with incubators and environmental chambers. Can be used in temperatures up to 40°C and humidity up to 80%. Supplied with a non-slip mat.
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)