Key Documents
Z651575
Quickfit® Pyrex® jacketed beakers
capacity 1,000 mL
About This Item
UNSPSC Code:
41121803
Pricing and availability is not currently available.
material
borosilicate glass
manufacturer/tradename
Quickfit 1025/22/SC
availability
available only in EU
capacity
1,000 mL
General description
Main part of the connectors are made from chemically resistant PTFE they can be used with temperature controlling mediums other than water, for example, oils. They are also suitable for use at temperatures from -20°C up to 200°C.
Other Notes
Browse all glass beakers
Legal Information
PYREX is a registered trademark of Corning, Inc.
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)