Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z650110

3M absorbents

P500 particulate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NB.35
Pricing and availability is not currently available.

manufacturer/tradename

P500 particulate

availability

available only in EU

absorption range measuring range

54 L

color

yellow

Looking for similar products? Visit Product Comparison Guide

General description

For control of spills of hazardous liquids. 3M Chemical Sorbents are made from inert, synthetic fibres principally of polypropylene. The have a high absorption capacity which minimises the amount of waste for disposal.
For fast coverage and absorption of spills.

Legal Information

3M is a trademark of 3M Company

pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service