Z645907
FORTUNA OPTIFIX® LOOP bottletop dispenser
20-100 mL, Solvent model with PTFE circulation system
All Photos(1)
About This Item
UNSPSC Code:
41121500
feature
graduations 2.0
with recirculating valve
manufacturer/tradename
Fortuna Graf 101.081-51H
Poulton & Graf 101.081-51H
measuring range
20-100 mL
volume
20-100 mL
fitting
GL 32
General description
The OPTIFIX LOOP is a patented and safe circulation system to fill and empty the bottle top dispenser. The benefit of this system is a unique, specialised dispensing nozzle which can circulate the dispensed chemical and therefore fill and empty the dispenser without waste. In closed position the LOOP system prevents aggressive liquid from coming out. The circulation system is made of solid PTFE. As the whole OPTIFIX bottletop dispenser does not contain any metal parts or seals, it is autoclavable. Accredited to ISO 8655.
Contents: 1 dispenser with 2 PP adapters(40&45) and 1 PTFE intake valve.
Basic dispenser model
Piston Type: PTFE-encased glass
Applications/features: Aqueous media, dilute acids, and NaOH. Has glass dispensing barrel.
Solvents dispenser model
Piston Type: Glass
Applications/features: Solvents and organics. Has safety-coated glass dispensing barrel. Recommended for dispensing deuterated NMR solvents packaged in 100 mL Aldrich Sure/Seal bottles.
Safety dispenser model
Piston Type: PTFE-encased glass
Applications/features: Strong or fuming acids and odoriferous products (e.g., bromine). Has extra lip seal at end of piston and PTFE/glass vent valve that prevents escape of vapors from bottle.
HF dispenser model
Piston Type: PTFE-encased glass
Applications/features: Hydrofluoric acid. Has extra lip seal at end of piston, PTFE dispensing cylinder, valve block, and PTFE/sapphire vent valve that prevents escape of vapors from bottle.
Contents: 1 dispenser with 2 PP adapters(40&45) and 1 PTFE intake valve.
Basic dispenser model
Piston Type: PTFE-encased glass
Applications/features: Aqueous media, dilute acids, and NaOH. Has glass dispensing barrel.
Solvents dispenser model
Piston Type: Glass
Applications/features: Solvents and organics. Has safety-coated glass dispensing barrel. Recommended for dispensing deuterated NMR solvents packaged in 100 mL Aldrich Sure/Seal bottles.
Safety dispenser model
Piston Type: PTFE-encased glass
Applications/features: Strong or fuming acids and odoriferous products (e.g., bromine). Has extra lip seal at end of piston and PTFE/glass vent valve that prevents escape of vapors from bottle.
HF dispenser model
Piston Type: PTFE-encased glass
Applications/features: Hydrofluoric acid. Has extra lip seal at end of piston, PTFE dispensing cylinder, valve block, and PTFE/sapphire vent valve that prevents escape of vapors from bottle.
made of PTFE with valve block made of PTFE incl. thread of GL 32 with PTFE encased dosing piston and PTFE cylinder
Legal Information
Optifix is a registered trademark of Poulten & Graf Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)