Z645737
Stuart™ melting point apparatus SMP10
AC/DC input 230 V AC
About This Item
Recommended Products
feature
CE compliant
Includes 100 melting point tubes
manufacturer/tradename
Bibby
temperature measuring range
≤300 °C, resolution: 0.1 °C (Accuracy ± 1.0 °C at 20 °C, ± 2.5 °C at 300 °C)
General description
Two samples can be tested simultaneously and viewed via a magnifying lens aided by built in illumination.
Supplied complete with calibration certificate.
- Digital selection and display of temperature
- Easy and safe to operate
- LED bulb
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)